Supelco® PESTANAL® Organic Reference Standard, Atrazine-2-Hydroxy
36631-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:2163-68-0
- Description:Atrazine-2-hydroxy
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00078693
- M-Clarity quality level:MQ100
- Synonym(s):2-Hydroxyatrazine
- Formula:C₈H₁₅N₅O
- Molecular weight:197,24
- Flash point:Not applicable