Saisie de commande
Belgium
ContactUsLinkComponent
VIC Dye qPCR Calibration Solution *10000X*
VIC Dye qPCR Calibration Solution *10000X*
Catalog # AATB67030
Supplier:  AAT Bioquest
New Product
CAS Number:  
undefined
VIC Dye qPCR Calibration Solution *10000X*
Catalog # AATB67030
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Cdt:
    100 µl
  • Désignation:
    VIC Dye qPCR calibration solution *10000X*
Specifications
About this item
VIC dye qPCR Calibration Solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ 7 systems.
  • Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Caution: For research use only.