ContactUsLinkComponent
HIS Lite™ Cy5 Tris NTA Chelator
HIS Lite™ Cy5 Tris NTA Chelator
Catalog # AATB12659
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
HIS Lite™ Cy5 Tris NTA Chelator
Catalog # AATB12659
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 µG
  • Description:
    HIS Lite™ Cy5 Tris NTA chelator
Specifications
About this item
Cy5-Tris NTA compound is a sensitive fluorescent chelator for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces with the addition of certain metal ions (such as Ni²⁺, Co²⁺ etc.) as a mediator.
  • Stable for 24 months
Quality tested in-house.
Caution: For research use only.