ContactUsLinkComponent
FluoroQuest™ TSA/PSA Antifade Mounting Medium *Optimised for Tyramide and Styramide Imaging*
Catalog # AATB44890
Supplier:  AAT Bioquest
New Product
CAS Number:  
undefined
FluoroQuest™ TSA/PSA Antifade Mounting Medium *Optimised for Tyramide and Styramide Imaging*
Catalog # AATB44890
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Pk:
    5 mL
  • Description:
    FluoroQuest™ TSA/PSA Antifade mounting medium *Optimised for Tyramide and Styramide imaging*
Specifications
About this item
FluoroQuest™ TSA/PSA Antifade mounting medium is the only mounting medium that is optimised for tyramide (TSA) and styramide (PSA)-based fluorescence imaging applications.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.