Order Entry
Belgium
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Isoproturon
Supelco® PESTANAL® Organic Reference Standard, Isoproturon
  36137-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Isoproturon
  36137-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    34123-59-6
  • Description:
    Isoproturon
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H351 - H361f - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078684
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-(4-Isopropylphenyl)-1,1-dimethylurea
  • Formula:
    C₁₂H₁₈N₂O
  • Molecular weight:
    206,28
  • Flash point:
    100 °C - closed cup

 

 

  • GHS signal: GHS08, GHS07, GHS09
 : P202 - P260 - P273 - P280 - P308 + P313 - P391