Order Entry
Belgium
ContactUsLinkComponent
Lithium 6 Internal Standard
Lithium 6 Internal Standard
Catalog # PROOVHGLISC6LI-100
New Product
CAS Number:  
Lithium 6 Internal Standard
Catalog # PROOVHGLISC6LI-100
New Product
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • GHS signal:
    Warning
  • Storage temperature:
    RT
  • Element:
    Lithium
  • Symbol:
    Li

Specifications

About this item