ContactUsLinkComponent
ReadiPrep™ Lipopolysaccharide (LPS) Isolation Kit
ReadiPrep™ Lipopolysaccharide (LPS) Isolation Kit
Catalog # AATB60201
Supplier:  AAT Bioquest
New Product
CAS Number:  
ReadiPrep™ Lipopolysaccharide (LPS) Isolation Kit
Catalog # AATB60201
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Pk:
    10
  • Pk Info:
    10 preparations
Specifications
About this item
Designed for consistent and dependable results. The ReadiPrep™ lipopolysaccharide (LPS) isolation kit provides a safe and effective way to isolate LPS from the outer membrane of Gram-negative bacteria.
  • Robust protocol
  • Unlike conventional approaches that rely on hazardous organic chemicals like chloroform or phenol, this kit provides a safer and more time-efficient alternative for isolating high-purity LPS for research applications
  • Six-month shelf life
Kit utilises a bacterial membrane lysis buffer and enzymatic protein digestion to extract pure LPS from bacterial cultures, which can be quantified using carbohydrate detection methods.
Delivery: Shipped with blue ice.
Caution: Research use only.