ContactUsLinkComponent
Amplite® Rapid Colorimetric PE-Maleimide Quantitation Kit
Amplite® Rapid Colorimetric PE-Maleimide Quantitation Kit
  AATB5530
New Product
 :  
Amplite® Rapid Colorimetric PE-Maleimide Quantitation Kit
  AATB5530
New Product
 :  
  • Pk:
    2 Tests
Designed for consistent and dependable results. The Amplite® rapid colorimetric PE-maleimide quantitation kit facilitates the quantification of maleimide groups on maleimide-activated PE proteins using a proprietary sensor, Maleimide 680™, which has an absorption peak at ~680 nm.
  • Robust protocol
  • This quantitation kit is compatible with various detection systems, including traditional cuvettes, NanoDrop™ Spectrophotometers, and 96-well absorbance plate readers
  • Six-month shelf life
It offers a robust and adaptable approach for the rapid quantification of maleimide modifications on PE proteins, addressing a critical need in protein biochemistry.
 : Shipped with blue ice.
 : Research use only.