Amplite® Fluorimetric Glucose and Sucrose Assay Kit
Catalog # AATB40010
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Pk:100 Tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® fluorimetric glucose and sucrose assay kit offers a convenient method for measuring glucose and sucrose levels in various biological samples, such as serum, plasma, body fluids, food, tissue extracts, and growth medium.
- Useful for assessing aerobic glycolysis, pyruvate kinase activity, and metabolic profiles in different biological matrices.
- Ex/Em = 571/584 nm
It is also employed for determining protein and sugar concentrations in food samples, as well as monitoring plasma insulin and glucose levels in murine models.
Delivery information: Shipped with blue ice.
Caution: Research use only.