ContactUsLinkComponent
Live or Dead™ Fixable Dead Cell Staining Kit
Live or Dead™ Fixable Dead Cell Staining Kit
Catalog # AATB22598
Fournisseur:  AAT Bioquest
New Product
Numéro CAS:  
Live or Dead™ Fixable Dead Cell Staining Kit
Catalog # AATB22598
Fournisseur:  AAT Bioquest
New Product
Numéro CAS:  
Spécifications
  • Pk:
    200 Tests
Spécifications
À propos de cet article
IR Fluorescence. Our Live or Dead™ fixable dead cell staining kits are a set of tools for labelling cells for fluorescence microscopic investigations of cellular functions.
  • The kit provides all the essential components with an optimized cell-labelling protocol
  • The fluorescent dye used in the kit is well excited with the IR laser (808 nm) and emits fluorescence at 878 nm (885/40 bandpass filter).
This particular kit is designed to uniformly label fixed mammalian cells in infrared fluorescence for flow cytometry applications with IR laser excitation.
Livraison: Shipped at RT.
Attention: Research use only.