ContactUsLinkComponent
Amplite® Fluorimetric Biotin Quantitation Kit
Amplite® Fluorimetric Biotin Quantitation Kit
Catalog # AATB5538
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Amplite® Fluorimetric Biotin Quantitation Kit
Catalog # AATB5538
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Tests per kit:
    100 tests
Specifications
About this item
Designed for consistent and dependable results. Amplite® Fluorimetric Biotin Quantitation Kit uses Biotinylite™ Green, a fluorogenic biotin sensor.
  • Robust protocol
  • Consistent results
  • 12-month shelf life
This fluorescence-based assay is much more sensitive than the commonly used colorimetric HABA assay.
Delivery: Shipped with blue ice.
Caution: Research use only.