ContactUsLinkComponent
Beadlite™ Rapid Colorimetric Thiol Quantitation Kit for Nanoparticles
Beadlite™ Rapid Colorimetric Thiol Quantitation Kit for Nanoparticles
Catalog # AATB5535
Supplier:  AAT Bioquest
New Product
CAS Number:  
Beadlite™ Rapid Colorimetric Thiol Quantitation Kit for Nanoparticles
Catalog # AATB5535
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Tests per kit:
    100 tests
Specifications
About this item
Designed for consistent and dependable results. Beadlite™ Rapid Colorimetric Thiol Quantitation kit provides a robust method to quantify thiol-functionalised nanoparticle with our proprietary Thiolite™ 490 that has its maximum absorbance at ~490 nm.
  • Robust protocol
  • Can be performed in a traditional cuvette, NanoDrop™ Spectrophotometer or a convenient 96-well absorbance plate reader
  • 12-month shelf life
This kit has been widely used for the rapid quantification of sulfhydryl groups on the surface of latex beads, agarose beads, or magnetic nanoparticles.
Delivery information: Shipped with blue ice.
Caution: Research use only.