ContactUsLinkComponent
Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit
Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit
  AATB5527
New Product
 :  
Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit
  AATB5527
New Product
 :  
  • Pk:
    2 Tests
  • Tests per kit:
    2 tests
Designed for consistent and dependable results. The Amplite® Rapid Colorimetric APC-Maleimide Quantitation Kit facilitates the quantification of maleimide groups on maleimide-activated APC proteins using a proprietary sensor, Maleimide 555™, which has an absorption peak at ~555 nm.
  • Robust protocol
  • This quantitation kit is compatible with various detection systems, including traditional cuvettes, NanoDrop™ Spectrophotometers, and 96-well absorbance plate readers
  • Six-month shelf life
Maleimide-based crosslinking reagents are commonly used for conjugating proteins to other proteins or biomolecules.
 : Shipped with blue ice.
 : Research use only.