ContactUsLinkComponent
Amplite® Colorimetric Hexokinase Assay Kit
Amplite® Colorimetric Hexokinase Assay Kit
Catalog # AATB11319
Supplier:  AAT Bioquest
New Product
CAS Number:  
Amplite® Colorimetric Hexokinase Assay Kit
Catalog # AATB11319
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Tests per kit:
    100 tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® Colorimetric Hexokinase Assay Kit is designed to accurately measure the activity of hexokinases from various sources using a simple and quick protocol.
  • Robust protocol
  • Absorbance measured at 450 nm
  • Six month shelf life
This kit utilises a coupled enzyme assay, where glucose is phosphorylated to glucose-6-phosphate and subsequently oxidised to produce NADH.
Delivery: Shipped with blue ice.
Caution: Research use only.