Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Dextran
Supelco® Organic Reference Standard, Dextran
Supplier:  Merck
Certificates
Supelco® Organic Reference Standard, Dextran
Supplier:  Merck

Specifications

  • CAS No.:
    9004-54-0
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00130935
  • M-Clarity quality level:
    MQ100
  • Formula:
    [C₆H₁₀O₅]ₙ

Specifications

Frequently Bought Together