Order Entry
Belgium
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, Dextran

Supelco® Organic Reference Standard, Dextran

Dextran, Analytical standard, for GPC, 80000, 100 mg
Catalog # 00892-100MG
Supplier:   Merck
Supelco® Organic Reference Standard, Dextran
Catalog # 00892-100MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Dextran

Dextran, Analytical standard, for GPC, 80000, 100 mg

CAS Number:  
Price
266,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard, for GPC, 80000
  • Description:
    Dextran
  • Cat. No.:
    00892-100MG
  • M-Clarity quality level:
    MQ100
  • Formula:
    [C₆H₁₀O₅]ₙ
  • Storage conditions:
    2...30 °C
  • MDL No.:
    MFCD00130935
  • Flash point:
    Not applicable
  • CAS No.:
    9004-54-0
  • Shipping conditions:
    STD

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.