Supelco® TraceCERT® Organic Reference Standard, Triadimenol
Triadimenol, 50 mg
Catalog # 38878-50MG
Supplier: Merck
Product Details & Documents
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P202 - P260 - P263 - P273 - P202 - P260 - P263 - P273 - P301 + P312 - P308 + P313 - P301 + P312 - P308 + P313
Specifications
Specifications
- Pk:50 mg
- Description:Triadimenol
- Cat. No.:38878-50MG
- M-Clarity quality level:MQ300
- Storage conditions:2...8 °C
- Hazard class description:H302 - H360D - H362 - H411 - H302 - H360D - H362 - H411
- MDL No.:MFCD00055507
- Synonym(s):α-tert-Butyl-β-(4-chlorophenoxy)-1H-1,2,4-triazole-1-ethanol
- Molecular weight:295,76
- CAS No.:55219-65-3
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



