Order Entry
Belgium
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, (−)-Guaiol
  29242-5MG
 :  Merck
 :  
Supelco® Organic Reference Standard, (−)-Guaiol
  29242-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    489-86-1
  • Description:
    (−)-Guaiol
  • Storage conditions:
    2...8 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00043336
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (3R,6S,10S)-6,10,α,α-Tetramethylbicyclo[5.3.0]dec-1(7)-ene-3-methanol
  • Melting point:
    91...93 °C (lit.); 91...93 °C
  • Molecular weight:
    222,37