ContactUsLinkComponent
HIS Lite™ Cy5 Tris NTA-Ni Complex
HIS Lite™ Cy5 Tris NTA-Ni Complex
Catalog # AATB12619
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
HIS Lite™ Cy5 Tris NTA-Ni Complex
Catalog # AATB12619
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 µG
  • Description:
    HIS Lite™ Cy5 tris NTA-Ni complex
Specifications
About this item
Cy5-Tris NTA compound is used as a sensitive fluorescent probe for detecting polyhistidine-labelled proteins in cells, solution and solid surfaces.
  • Stable for 12 months
Quality tested in-house.
Caution: For research use only.