Order Entry
Belgium
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Trichlorfon
Supelco® TraceCERT® Organic Reference Standard, Trichlorfon
  CRM67618-50MG
 :  Merck
 :  
Supelco® TraceCERT® Organic Reference Standard, Trichlorfon
  CRM67618-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    52-68-6
  • Description:
    Trichlorfon
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 + H312 - H317 - H334 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055272
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    (2,2,2-Trichloro-1-hydroxyethyl)phosphonic acid dimethyl ester; Metrifonate
  • Molecular weight:
    257,44

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
 : P261 - P273 - P280 - P301 + P312 - P302 + P352 + P312 - P304 + P340 + P312