Supelco® TraceCERT® Organic Reference Standard, 1-Naphthylacetamide
Catalog # CRM43557-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:86-86-2
- Description:1-Naphthylacetamide
- Storage conditions:15...25 °C
- Hazard class description:H302 - H318 - H361d - H412
- Shipping conditions:STD
- MDL No.:MFCD00004047
- M-Clarity quality level:MQ300
- Synonym(s):1-Naphthaleneacetamide
- Melting point:180 - 183 °C (lit.)
- Formula:C₁₀H₇CH₂CONH₂
- Molecular weight:185,22
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P201 - P273 - P280 - P301 + P312 - P305 + P351 + P338 - P308 + P313