Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Farnesol
Supelco® Organic Reference Standard, Farnesol
Catalog # 93547-100MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Farnesol
Catalog # 93547-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Mixture of isomers, analytical standard
  • CAS No.:
    4602-84-0
  • Description:
    Farnesol
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H315 - H317 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002918
  • Synonym(s):
    3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol
  • Formula:
    (CH₃)₂C=CHCH₂CH₂C(CH₃)=CHCH₂CH₂C(CH₃)=CHCH₂OH
  • Molecular weight:
    222,37
  • Density:
    0,886 g/ml at 20 °C (lit.)
  • Flash point:
    155 °C - Closed cup
  • Boiling point:
    149 °C/4 mmHg (lit.)

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P261 - P264 - P272 - P273 - P280 - P302 + P352