Supelco® Organic Reference Standard, L-(−)-Fucose
Catalog # 93183-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:2438-80-4
- Description:L-(−)-Fucose
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00135607
- M-Clarity quality level:MQ100
- Synonym(s):6-Deoxy-ʟ-galactose
- Melting point:150 - 153 °C (lit.)
- Molecular weight:164,16