Supelco® Organic Reference Standard, 2-Hydroxy-3-methylvaleric acid
Catalog # 80529-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Mixture of diastereomers, analytical standard
- CAS No.:488-15-3
- Description:2-Hydroxy-3-methylvaleric acid
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD14624640
- M-Clarity quality level:MQ100
- Synonym(s):2-Hydroxy-3-methylpentanoic acid
- Molecular weight:132,16