Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Biotin azide
Biotin azide, FastClick™
Biotin azide, FastClick™
Biotin azide, FastClick™
Catalog # AATB72801
Fournisseur:
AAT Bioquest
New Product
Numéro CAS:
Expand image
Biotin azide, FastClick™
Catalog # AATB72801
Fournisseur:
AAT Bioquest
New Product
Numéro CAS:
Spécifications
Pk:
1 mg
Seq:
2070
Spécifications
À propos de cet article
Product Details
:
:
326.42 g/mol
Product Details