Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cy3 phosphoramidite
Cy3 phosphoramidite
Cy3 phosphoramidite
Cy3 phosphoramidite
Catalog # AATB6070
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
Cy3 phosphoramidite
Catalog # AATB6070
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
100 µmol
Seq:
9140
Specifications
About this item
Product Details
:
:
953.63 g/mol
CAS Number:
182873-76-3
Product Details