Anti-beta Catenin Rabbit Recombinant Antibody [clone: CTNNB1/7044R]
Catalog # ANTIA277968-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:beta Catenin
- Antigen symbol:beta Catenin
- Clonality:Recombinant
- Clone:CTNNB1/7044R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P35222
- Antigen synonyms:CTNB1_HUMAN|CHBCAT|CATNB|Catenin beta 1|Catenin (cadherin associated protein), beta 1, 88kDa|Beta-catenin|Catenin beta-1|CTNNB|Cadherin associated protein
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:92 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human beta Catenin protein.
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [CTNNB1/7044R] antibody to beta Catenin for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: beta Catenin
Clonality: Recombinant
Clone: CTNNB1/7044R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human