Order Entry
Belgium
ContactUsLinkComponent
Anti-Wilms Tumor Protein Mouse Recombinant Antibody [clone: rWT1/6908]
Anti-Wilms Tumor Protein Mouse Recombinant Antibody [clone: rWT1/6908]
Catalog # ANTIA277929-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Wilms Tumor Protein Mouse Recombinant Antibody [clone: rWT1/6908]
Catalog # ANTIA277929-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AWT1
  • Antigen symbol:
    Wilms Tumor Protein
  • Clonality:
    Recombinant
  • Clone:
    rWT1/6908
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P19544
  • Antigen synonyms:
    Wilms Tumor Protein|WAGR|FWT1|NPHS4|Wilms' tumor protein|Wilms Tumor|GUD|Wilms' tumor gene|Wilms tumor 1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    49 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 1-100, of human Wilms Tumor Protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant mouse monoclonal [rWT1/6908] antibody to Wilms Tumor Protein for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Wilms Tumor Protein
Clonality: Recombinant
Clone: rWT1/6908
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human