Order Entry
Belgium
ContactUsLinkComponent
Anti-Desmin Mouse Recombinant Antibody [clone: rDES/1711]
Anti-Desmin Mouse Recombinant Antibody [clone: rDES/1711]
Catalog # ANTIA277883-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Desmin Mouse Recombinant Antibody [clone: rDES/1711]
Catalog # ANTIA277883-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CMD1I
  • Antigen symbol:
    Desmin
  • Clonality:
    Recombinant
  • Clone:
    rDES/1711
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Hamster,
    Chicken,
    Human,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P17661
  • Antigen synonyms:
    CSM1|FLJ41013|DESM_HUMAN|DES|Desmin|FLJ12025|FLJ39719|FLJ41793|CSM2
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    52 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human Desmin protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant mouse monoclonal [rDES/1711] antibody to Desmin for IHC-P with samples derived from human, mouse, rat, hamster and chicken.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Desmin
Clonality: Recombinant
Clone: rDES/1711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat;Hamster;Chicken