Anti-Wilms Tumor Protein Mouse Monoclonal Antibody [clone: 6F-H2]
Catalog # ANTIA253510-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AWT1
- Antigen symbol:Wilms Tumor Protein
- Clonality:Monoclonal
- Clone:6F-H2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat
- Form:Liquid
- Gene ID:UniprotID# P19544
- Antigen synonyms:Wilms Tumor Protein|WAGR|FWT1|NPHS4|Wilms' tumor protein|Wilms Tumor|GUD|Wilms' tumor gene|Wilms tumor 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:47 - 55 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, corresponding to amino acids 1-181, of human WT1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [6F-H2] antibody to Wilms Tumor Protein for IHC-P with samples derived from human and rat.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Wilms Tumor Protein
Clonality: Monoclonal
Clone: 6F-H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat