Anti-Wilms Tumor Protein Mouse Monoclonal Antibody [clone: ABT-WT1]
Supplier: Antibodies.com
New Product
Certificates
About this item
Mouse monoclonal (ABT-WT1) antibody to Wilms' Tumor 1 for IHC with samples derived from Human.
- Validated Applications: IHC
Type: Primary
Antigen: Wilms Tumor Protein
Clonality: Monoclonal
Clone: ABT-WT1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ANTIA97062-100
- ANTIA97062-50
- Antigen symbol:
- Wilms Tumor Protein
- Wilms Tumor Protein
- Antigen name:
- AWT1
- AWT1
- Antigen synonyms:
- Wilms Tumor Protein|WAGR|FWT1|NPHS4|Wilms' tumor protein|Wilms Tumor|GUD|Wilms' tumor gene|Wilms tumor 1
- Wilms Tumor Protein|WAGR|FWT1|NPHS4|Wilms' tumor protein|Wilms Tumor|GUD|Wilms' tumor gene|Wilms tumor 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-WT1
- ABT-WT1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P19544
- UniprotID# P19544
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human Wilms' Tumor 1
- Synthetic peptide derived from human Wilms' Tumor 1
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage buffer:
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- IHC
- IHC
Specifications
Frequently Bought Together