Anti-CREB3L2 Rabbit Polyclonal Antibody
Plastic vial
Catalog # ANTIA96583-100
Supplier: Antibodies.com
Jump to:
Product Details & Documents
Rabbit polyclonal antibody to CREB3L2 for WB and ELISA with samples derived from Human, Mouse and Rat.
Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000.
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: CREB3L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Seq:1286
- Pack type:Plastic vial
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Molecular weight:57 kDa
- Shipping temperature:Shipped on blue ice at 4 °C
- Antigen name:B ZIB transcription factor
- Cat. No.:ANTIA96583-100
- Reactivity:Human|Mouse|Rat
- Gene ID:UniprotID# Q70SY1
- Antigen synonyms:BBF2 human homolog on chromosome 7|BBF2H7|cAMP-responsive element-binding protein 3-like protein 2|Cyclic AMP-responsive element-binding protein 3-like protein 2
- Immunogen:Synthetic peptide derived from human CREB3L2 (amino acids 269-318)
- Conjugation:Unconjugated
- Antigen symbol:CREB3L2
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- ELISA:Yes



