Order Entry
Belgium
ContactUsLinkComponent
 
Anti-GST3/GST pi Rabbit Monoclonal Antibody [clone: ARC0439]
  ANTIA306698-100
New Product
undefined
Anti-GST3/GST pi Rabbit Monoclonal Antibody [clone: ARC0439]
  ANTIA306698-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Deafness
  • Antigen symbol:
    GST3 / GST pi
  • Clonality:
    Monoclonal
  • Clone:
    ARC0439
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P09211
  • Antigen synonyms:
    GST class-pi|Glutathione S-transferase pi 1|Glutathione S-transferase P|Fatty Acid Ethyl Ester Synthase III|Glutathione S Transferase 3|Glutathione S Transferase Pi|Deafness X-linked 7|DFN7|FAEES3
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    25 kDa
  • Sequence:
    LRCKYISLIYTNYEAGKDDYVKALPGQLKPFETLLSQNQGGKTFIVGDQISFADYNLLDLLLIHEVLAPGCLDAFPLLSAYVGRLSARPKLKAFLASPEYVN
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 100-201 of human GST3/GSTP1 (P09211).
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit monoclonal [ARC0439] antibody to GST3/GST pi for WB and IHC with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type: Primary
Antigen: GST3 / GST pi
Clonality: Monoclonal
Clone: ARC0439
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat