Order Entry
Belgium
ContactUsLinkComponent
Anti-HAO2 Goat Polyclonal Antibody
Anti-HAO2 Goat Polyclonal Antibody
Catalog # ANTIA83649-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-HAO2 Goat Polyclonal Antibody
Catalog # ANTIA83649-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    (S)-2-hydroxy-acid oxidase
  • Antigen symbol:
    HAO2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    , peroxisomal|Glycolate oxidase|HAOX2|2 hydroxy acid oxidase peroxisomal|HAO2|GIG16|HAO 2|Cell growth-inhibiting gene 16 protein|HAOX2_HUMAN
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
  • Sequence:
    C-DVSEVDTRTTIQGE
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human HAO2 (internal region)
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Goat polyclonal antibody to HAO2 for ELISA and WB with samples derived from human.

  • Validated applications: ELISA, WB

Type: Primary
Antigen: HAO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human