ContactUsLinkComponent
ReadiUse™ Universal Kinase Reaction Stopping Buffer
ReadiUse™ Universal Kinase Reaction Stopping Buffer
Catalog # AATB31005
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
ReadiUse™ Universal Kinase Reaction Stopping Buffer
Catalog # AATB31005
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    ReadiUse™ Universal kinase reaction stopping buffer
  • Environmentally Preferable:
Specifications
About this item
ReadiUse™ Universal Kinase Reaction Stopping Buffer is an aqueous solution that effectively chelates divalent metal ions.
  • Designed for ease of use
  • Saves time
This sterile-filtered solution finds wide applicability in biochemistry and molecular biology research.
Caution: For research use only.