ContactUsLinkComponent
Fluorescein-12-dCTP *1 mM in Tris Buffer (pH 7,5)*
Fluorescein-12-dCTP *1 mM in Tris Buffer (pH 7,5)*
Catalog # AATB17035
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
Fluorescein-12-dCTP *1 mM in Tris Buffer (pH 7,5)*
Catalog # AATB17035
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    25 Nmo
Specifications
About this item
Fluorescein-12-dCTP might be incorporated into a DNA sequence as its natural counterpart dCTP to incorporate a fluorescein tag in a DNA molecule.
  • Can be used for the direct enzymatic labelling of DNA/cDNA, e.g., by PCR or nick translation
The linker between fluorescein tag and cytidine has been optimised for the optimal substrate incorporation and labelling efficiency.
Caution: For research use only.