Anti-Wilms Tumor Protein Rabbit Recombinant Antibody [clone: WT1/1434R]
Catalog # ANTIA250335-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AWT1
- Antigen symbol:Wilms Tumor Protein
- Clonality:Recombinant
- Clone:WT1/1434R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P19544
- Antigen synonyms:Wilms Tumor Protein|WAGR|FWT1|NPHS4|Wilms' tumor protein|Wilms Tumor|GUD|Wilms' tumor gene|Wilms tumor 1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:47 - 55 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human WT1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [WT1/1434R] antibody to Wilms Tumor Protein for IHC-P with samples derived from Human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Wilms Tumor Protein
Clonality: Recombinant
Clone: WT1/1434R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human