Anti-MBD3 Mouse Monoclonal Antibody [clone: PCRP-MBD3-1C4]
ANTIA252819-100
New Product
- Antibody type:Primary
- Antigen name:AI181826
- Antigen symbol:MBD3
- Clonality:Monoclonal
- Clone:PCRP-MBD3-1C4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# O95983
- Antigen synonyms:MBD3|Methyl CpG binding protein MBD3|MBD3: methyl CpG binding domain protein 3|Methyl-CpG-binding protein MBD3|MBD 3|Methyl-CpG-binding domain protein 3|AU019209|MBD3_HUMAN
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:34 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human MBD3 protein
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PCRP-MBD3-1C4] antibody to MBD3 for Flow Cytometry and IF with samples derived from Human.
- Validated applications: Flow cytometry, IF
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: MBD3
Clonality: Monoclonal
Clone: PCRP-MBD3-1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human