Order Entry
Belgium
ContactUsLinkComponent
 
Anti-Wilms Tumor Protein Mouse Monoclonal Antibody [clone: 6F-H2]
  ANTIA250330-100
New Product
undefined
Anti-Wilms Tumor Protein Mouse Monoclonal Antibody [clone: 6F-H2]
  ANTIA250330-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    AWT1
  • Antigen symbol:
    Wilms Tumor Protein
  • Clonality:
    Monoclonal
  • Clone:
    6F-H2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P19544
  • Antigen synonyms:
    Wilms Tumor Protein|WAGR|FWT1|NPHS4|Wilms' tumor protein|Wilms Tumor|GUD|Wilms' tumor gene|Wilms tumor 1
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    47 - 55 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, corresponding to amino acids 1 - 181, of human WT1 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [6F-H2] antibody to Wilms Tumor Protein for IHC-P with samples derived from Human and Rat.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Wilms Tumor Protein
Clonality: Monoclonal
Clone: 6F-H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat