Anti-NAPSIN A Mouse Recombinant Antibody [clone: rNAPSA/7239]
ANTIA316747-100
New Product
- Antibody type:Primary
- Antigen name:Asp 4
- Antigen symbol:NAPSIN A
- Clonality:Recombinant
- Clone:rNAPSA/7239
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# O96009
- Antigen synonyms:Kidney derived aspartic protease like protein|NAPA|Kdap|Aspartyl protease 4|KAP|NAP1|Napsa|NAPSA_HUMAN|ASP4
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:37 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (aa50-150) of human Napsin A protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant mouse monoclonal [rNAPSA/7239] antibody to NAPSIN A for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: NAPSIN A
Clonality: Recombinant
Clone: rNAPSA/7239
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human