ContactUsLinkComponent
ReadiUse™ ATP Reference Standard *10 mM Aqueous Solution*
ReadiUse™ ATP Reference Standard *10 mM Aqueous Solution*
Catalog # AATB17198
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
ReadiUse™ ATP Reference Standard *10 mM Aqueous Solution*
Catalog # AATB17198
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    5 mL
  • Description:
    ReadiUse™ ATP Reference Standard 10 mM aqueous solution
Specifications
About this item
The ReadiUse™ ATP Reference Standard *10 mM Aqueous Solution* is designed for the accurate quantification of adenosine triphosphate (ATP) in various scientific assays.
  • High purity
  • Supplied in an aqueous solution for immediate use in calibrating ATP measurement assays
The ReadiUse™ ATP Reference Standard is applicable for various scientific purposes, including calibration of bioluminescence assays for ATP detection, assessment of microbial contamination, evaluation of cell viability and proliferation, investigation of energy metabolism pathways and environmental monitoring for microbial load.
Caution: For research use only.