ContactUsLinkComponent
DEAC-dUTP *1 mM in TE Buffer (pH 7,5)*
DEAC-dUTP *1 mM in TE Buffer (pH 7,5)*
Catalog # AATB17034
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
DEAC-dUTP *1 mM in TE Buffer (pH 7,5)*
Catalog # AATB17034
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 Nmo
Specifications
About this item
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling or PCR.
  • Chemically identical to Diethylaminocoumarin-5-dUTP (Perkin Elmer’s NEL455001EA)
This Coumarin-dUTP conjugate can be used as a blue fluorescence colour with Spectrum Aqua™ filter set (Spectrum Aqua™ is the trademark of Vysis).
Caution: For research use only.