ContactUsLinkComponent
iFluor® 560 dUTP *1 mM in TE Buffer (pH 7,5)*
iFluor® 560 dUTP *1 mM in TE Buffer (pH 7,5)*
Catalog # AATB17033
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
iFluor® 560 dUTP *1 mM in TE Buffer (pH 7,5)*
Catalog # AATB17033
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    25 Nmo
Specifications
About this item
iFluor® 560-dUTP is a iFluor 560-modified nucleotide that can be used in various molecular biology applications, such as labelling and detection of DNA molecules.
  • iFluor® 560-dUTP is preferred over other similar fluorescent dyes due to its high fluorescence intensity, photostability and compatibility with a wide range of detection systems
  • Used in techniques such as fluorescence in situ hybridisation (FISH), microarray analysis and real-time PCR
When incorporated into DNA during synthesis, the iFluor® 560-dUTP can be visualized using fluorescence microscopy or other fluorescence-based assays.
Caution: For research use only.