Order Entry
Belgium
ContactUsLinkComponent
Anti-NMDAR2A Mouse Monoclonal Antibody [Clone: S327-95]
Anti-NMDAR2A Mouse Monoclonal Antibody [Clone: S327-95]
  ANTIA304974-100
New Product
undefined
Anti-NMDAR2A Mouse Monoclonal Antibody [Clone: S327-95]
  ANTIA304974-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    EPND
  • Antigen symbol:
    NMDAR2A
  • Clonality:
    Monoclonal
  • Clone:
    S327-95
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q00959
  • Antigen synonyms:
    hNR2A|LKS|GRIN2A|GluN2A|GRIN 2A|Glutamate [NMDA] receptor subunit epsilon-1|FESD|Glutamate receptor|Glutamate receptor ionotropic N methyl D aspartate 2A
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    ~170 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Fusion protein amino acids 75 - 325 (extracellular N - terminus) of rat GluN2A/NR2A.
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Protein G purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [S327-95] antibody to NMDAR2A for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:1000

Type: Primary
Antigen: NMDAR2A
Clonality: Monoclonal
Clone: S327-95
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat