Phenol
Black HDPE can, 1 L
Catalog # PRLS90341.290
Supplier: PROLABO CMP
Jump to:
Product Details & Documents
In accordance with the operating mode: 50 mg SODIUM PENTACYANONITROSYLFERRATE 2H2O ANALAR Ref VWR 27966 10 g PHENOL FOR ACS ANALYSES Ref Merck 100206 QSP 1000 ml deionised water.
Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene
- CAS Number:108-95-2
- Formula:C₆H₅OH
- MDL Number:MFCD00002143
- Molecular Weight:94.11 g/mol
- Boiling Pt:100 °C
- Density:1 g/cm³
Specifications
Specifications
- Pack type:Black HDPE can
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



