Order Entry
Belgium
Orders LinkContactUsLinkComponent
Nitric acid

Nitric acid

Colourless HDPE can, 10 L
Catalog # PRLS90335.9010
Toxic
Corrosive
Supplier:   PROLABO CMP
Nitric acid
Catalog # PRLS90335.9010
Supplier:  PROLABO CMP
CAS Number:  7697-37-2
Nitric acid

Colourless HDPE can, 10 L

CAS Number:  7697-37-2
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents

In accordance with the operating mode: 7,5 liters nitric acid 69% Ref VWR 20425 2,5 liters deionised water.

Synonyms:  Nitric acid solution
  • CAS Number:7697-37-2
  • Formula:HNO₃
  • MDL Number:MFCD00011349
  • Molecular Weight:63.01 g/mol
  • Boiling Pt:100 °C
  • UN:2031
  • ADR:8,II
  •  :Density: 1.51 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Colourless HDPE can
  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.