Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Hydrochloric acid

Colourless HDPE can, 30 L
Catalog # PRLS90333.9030
Corrosive
Irritant
Supplier:   PROLABO CMP
Hydrochloric acid
Catalog # PRLS90333.9030
Supplier:  PROLABO CMP
CAS Number:  7647-01-0
Hydrochloric acid

Colourless HDPE can, 30 L

CAS Number:  7647-01-0
Price
Inquire For Price
Log in for availability
1 * 30 L
Product Details & Documents

30 liters Hydrochloric acid 35% RECTAPUR Ref VWR 20248.

Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:85 °C
  • Density:1,18 g/cm³
  • UN:1789
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Colourless HDPE can
  • Pk:
    30 L

Specifications