Specifications
- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Aspartate beta Hydroxylase
- Description:Human Aspartate beta Hydroxylase ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:20 - 320 ng/L
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:1,24 ng/L
- Regulatory status:RUO
Specifications
About this item
Human Aspartate beta Hydroxylase ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Aspartate beta Hydroxylase in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human Aspartate beta Hydroxylase ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Aspartate beta Hydroxylase in serum, plasma or other biological fluids. An antibody specific for Aspartate beta Hydroxylase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Aspartate beta Hydroxylase present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Aspartate beta Hydroxylase antibody, which also binds the Aspartate beta Hydroxylase present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Aspartate beta Hydroxylase antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Aspartate beta Hydroxylase antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of Aspartate beta Hydroxylase captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Aspartate beta Hydroxylase in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.