Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Mouse RSPO1 ELISA Kit

Mouse RSPO1 ELISA kit, 96 Tests
  ANTIA313394-96
Mouse RSPO1 ELISA Kit
  ANTIA313394-96
 :  
Mouse RSPO1 ELISA Kit
Mouse RSPO1 ELISA Kit

Mouse RSPO1 ELISA kit, 96 Tests

 :  
 
565,00 €
   
1 * 96 Tests
 

Mouse RSPO1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse RSPO1 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 25 to 400 ng/L
  • Sensitivity: 0,98 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse RSPO1 ELISA kit (A313394) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse RSPO1 in serum, plasma or other biological fluids. An antibody specific for RSPO1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the RSPO1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-RSPO1 antibody, which also binds the RSPO1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-RSPO1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-RSPO1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of RSPO1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of RSPO1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

 

 

  • Pk:
    96 Tests
  • Description:
    Mouse RSPO1 ELISA kit
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma or other biological fluids
  • Sample volume:
    40 μl
  • Detection range:
    25 - 400 ng/L
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ANTIA313394-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,98 ng/L
  • Format:
    Pre-coated
  • Target protein:
    RSPO1
  • Regulatory status:
    RUO