Mouse Edn3 ELISA Kit
Product Details & Documents
Mouse Edn3 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Edn3 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 15 to 240 ng/L
- Sensitivity: 0,54 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Mouse Edn3 ELISA kit (A313573) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Edn3 in serum, plasma or other biological fluids. An antibody specific for Edn3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Edn3 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Edn3 antibody, which also binds the Edn3 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Edn3 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Edn3 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Edn3 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Edn3 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Specifications
Specifications
- Pk:96 Tests
- Description:Mouse Edn3 ELISA kit
- Primary antibody reactivity:Mouse
- Sample type:Serum, plasma or other biological fluids
- Sample volume:40 μl
- Detection range:15 - 240 ng/L
- Storage temperature:4 °C
- Shelf life:Store for 6 months at 4 °C
- Assay Type:Sandwich
- Time to Results:1 h 30 min
- Cat. No.:ANTIA313573-96
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:0,54 ng/L
- Format:Pre-coated
- Target protein:Edn3
- Regulatory status:RUO
Specifications
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



